missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P2X1 Monoclonal Antibody (551820)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560893

Product Code. 30535272

  • £332.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with relatively high calcium permeability. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle, being responsible, for example, for sympathetic vasoconstriction in small arteries, arterioles and vas deferens. Mouse studies suggest that this receptor is essential for normal male reproductive function. It is possible that the development of selective antagonists for this receptor may provide an effective non-hormonal male contraceptive pill.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2X1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P51575
P2RX1
Human embryonic kidney cell, HEK293-derived transfected with human P2X1/P2RX1.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry
551820
Unconjugated
P2RX1
AI323649; ATP receptor; BB122383; P2RX1; P2x; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; P2XMR; Pdcd3; programmed cell death 3; Purinergic receptor; purinergic receptor P2X 1; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; purinergic receptor P2X1-1; purinergic receptor P2X1-2; RP-2; RP-2 protein
Mouse
Protein A/G
RUO
5023
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.