missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GeneBLAzer™ P2RY2-NFAT-bla CHO-K1 Cells
Description
- Gene Symbol: P2RY2
- Accession Number: BC028135.1
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ P2RY2-NFAT-bla CHO-K1 cells contain the human Purinergic receptor P2, G protein-coupled, 2 (P2RY2) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
- The GeneBLAzer™ P2RY2-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50concentrations of ATP. In addition, GeneBLAzer™ P2RY2-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
- P2RY2 expression has been identified in smooth muscle, skeletal muscle, heart, spleen, lymphocytes, macrophages, bone marrow, lung, intestine, placenta, brain, kidney, and liver. P2RY2 has been implicated in the induction of chloride secretion in airway epithelial tissue, increases in coronary blood flow by vasodilation, and epidermal homeostasis.
Specifications
Specifications
| Content And Storage | Liquid Nitrogen |
| Detection Method | Fluorescence |
| For Use With (Application) | GPCRs |
| Includes | P2RY2-NFAT-bla CHO-K1 cells, P2RY2-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis |
| Quantity | 2 x 106 Cells |
| Type | Dividing Cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?