missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p27Kip1 Mouse, Clone: DCS-72.F6, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1027-MSM3-P1
This item is not returnable.
View return policy
Specifications
| p27Kip1 | |
| Nuclear | |
| DCS-72.F6 | |
| Unconjugated | |
| NP_004055 | |
| Mouse | |
| 25-26kDa | |
| Protein A/G Purified | |
| RUO | |
| 1027 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| CDKN1B | |
| Mouse recombinant p27 protein | |
| ZR75, T47D, SK-BR-3, MDA-MB-231, HeLa or MCF7 cells. Tonsil, Breast, Cervical or Colon Carcinoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction