missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552634
This item is not returnable.
View return policy
Description
Immunogen sequence: ITFELPDNAK QCFYEDIAQG TKCTLEFQVI TGGHYDVDCR LEDPDGKVLY KEMKKQYDSF TFTASKNGTY KFCFSNEFST F Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
p27, also known as p27Kip1, is a crucial cyclin-dependent kinase inhibitor (CKI) involved in the regulation of the cell cycle. The p27 protein is encoded by the CDKN1B gene located on chromosome 12 and plays a vital role in controlling cell proliferation by inhibiting cyclin-CDK complexes, particularly cyclin A-CDK2 and cyclin E-CDK2. By regulating these complexes, p27 induces cell cycle arrest at the G1 phase, preventing cells from prematurely entering the S phase. This function is critical for maintaining normal cellular processes and preventing uncontrolled cell division. p27 acts as a tumor suppressor, and its expression is tightly controlled by transcriptional and post-transcriptional mechanisms, including proteasome-mediated degradation. Reduced levels of p27 have been associated with numerous cancers, highlighting its role in tumorigenesis. The regulation of p27 involves signaling pathways such as the PI3K/AKT pathway, impacting its localization and stability in response to extracellular signals.
Specifications
| p27 | |
| Polyclonal | |
| Unconjugated | |
| TMED7 | |
| CGI-109; p24 family protein gamma-3; p24g3; p24gamma3; p27; TMED7; Transmembrane emp24 domain-containing protein 7; transmembrane emp24 protein transport domain containing 7; transmembrane p24 trafficking protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y3B3 | |
| TMED7 | |
| Recombinant protein corresponding to Human p27. Recombinant protein control fragment (Product #RP-90226). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction