missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p27/Kip1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £443.00
Specifications
| Antigen | p27/Kip1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214722
|
Novus Biologicals
NBP2-56105 |
100 μL |
£443.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625298
|
Novus Biologicals
NBP2-56105-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p27/Kip1 Polyclonal specifically detects p27/Kip1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| p27/Kip1 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Prostate Cancer, Tumor Suppressors | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 1027 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PAAPLIGAPANSEDTHLVDPKTDPSDSQTGLAEQCAGIRKRPATDDSSTQNKRANRTEENVSDGSPNAGSVEQTPKKPGLRRRQT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| CDKN1B | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title