missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p23 Recombinant Rabbit Monoclonal Antibody (10M9W8)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA538139

Product Code. 17257013

  • £463.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: Rat testis, Rat brain, HeLa, HepG2, Jurkat, Mouse testis, Mouse brain Immunogen sequence: MQPASAKWYD RRDYVFIEFC VEDSKDVNVN FEKSKLTFSC LGGSDNFKHL NEIDLFHCID PNDSKHKRTD RSILCCLRKG ESGQSWPRLT KERAKLNWLS.

Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Prior to activation, steroid receptors associate with a number of different proteins in both a stable and transient fashion. Steroid receptor complex proteins include heat shock proteins (HSP70 and HSP90), immunosuppressant binding proteins called immunophilins (the FK506 binding proteins, FKBP52 & FKBP54 and the cyclosporin binding protein, CyP-40) and at least three other proteins termed p23, p60 and p48. p23 along with HSP70, HSP90 and p60, combine with progesterone receptor (PR) as members of a transient intermediate complex. Cloned human p23 encodes a protein of 160 amino acids that is highly conserved between species and shows no homology to previously identified proteins. p23 is a highly acidic phosphoprotein with an aspartic acid-rich C-terminal domain and multiple potential phosphorylation sites. In vitro studies have suggested that p23 binds to HSP90 and is necessary for the binding of HSP90 and CyP-40 to PR. While neither its exact function nor mechanism of action have been identified, p23 appears to be an important factor in PR function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

p23
Recombinant Monoclonal
0.65 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P83868, Q15185, Q9R0Q7
Ptges3
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human P23/PTGES3 (NP_0065923).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot, Immunocytochemistry
10M9W8
Unconjugated
Ptges3
5730442A20Rik; Co chaperone p23; cPGES; cytosolic prostaglandin E synthase; cytosolic prostaglandin E2 synthase; Hsp90 co chaperone; Hsp90 co-chaperone; LOC100351480; P23; p23 cochaperone; progesterone receptor; progesterone receptor complex p23; Prostaglandin E synthase 3; prostaglandin E synthase 3 (cytosolic); prostaglandin-E synthase 3; Ptges; PTGES3; ptges3 {ECO:0000250; RGD1561913; sid 3177; Sid3177; Tebp; telomerase binding protein, p23; telomerase-binding protein p23; unactive progesterone receptor, 23 kD; UniProtKB:Q15185}
Rabbit
Affinity Chromatography
RUO
10728, 362809, 56351
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.