missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p23/PTGES3 Rabbit anti-Human, Mouse, Rat, Clone: 10M9W8, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16586-20UL
This item is not returnable.
View return policy
Description
p23/PTGES3 Monoclonal antibody specifically detects p23/PTGES3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| p23/PTGES3 | |
| Monoclonal | |
| Unconjugated | |
| cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human p23/PTGES3 (Q15185). | |
| 20 μg | |
| Membrane Trafficking and Chaperones | |
| 10728 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| 10M9W8 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction