Learn More
Invitrogen™ p23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529547
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain, RK3E cells. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (86%), Chicken (96%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Prior to activation, steroid receptors associate with a number of different proteins in both a stable and transient fashion. Steroid receptor complex proteins include heat shock proteins (HSP70 and HSP90), immunosuppressant binding proteins called immunophilins (the FK506 binding proteins, FKBP52 & FKBP54 and the cyclosporin binding protein, CyP-40) and at least three other proteins termed p23, p60 and p48. p23 along with HSP70, HSP90 and p60, combine with progesterone receptor (PR) as members of a transient intermediate complex. Cloned human p23 encodes a protein of 160 amino acids that is highly conserved between species and shows no homology to previously identified proteins. p23 is a highly acidic phosphoprotein with an aspartic acid-rich C-terminal domain and multiple potential phosphorylation sites. In vitro studies have suggested that p23 binds to HSP90 and is necessary for the binding of HSP90 and CyP-40 to PR. While neither its exact function nor mechanism of action have been identified, p23 appears to be an important factor in PR function.
Specifications
| p23 | |
| Polyclonal | |
| Unconjugated | |
| Ptges3 | |
| 5730442A20Rik; Co chaperone p23; cPGES; cytosolic prostaglandin E synthase; cytosolic prostaglandin E2 synthase; Hsp90 co chaperone; Hsp90 co-chaperone; LOC100351480; P23; p23 cochaperone; progesterone receptor; progesterone receptor complex p23; Prostaglandin E synthase 3; prostaglandin E synthase 3 (cytosolic); prostaglandin-E synthase 3; Ptges; PTGES3; ptges3 {ECO:0000250; RGD1561913; sid 3177; Sid3177; Tebp; telomerase binding protein, p23; telomerase-binding protein p23; unactive progesterone receptor, 23 kD; UniProtKB:Q15185} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10728, 362809, 56351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.31 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P83868, Q15185, Q9R0Q7 | |
| Ptges3 | |
| Full length human p23 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.