missing translation for 'onlineSavingsMsg'
Learn More

p21WAF1 Ab-6 Mouse Monoclonal Antibody, Epredia™

Achieve accurate, reproducible results with the Epredia™ p21WAF1 Ab-6, Mouse Monoclonal Antibody.

Brand:  Epredia MS-454-P0

 View more versions of this product

Product Code. 12651477

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: HJ21; same as WA-1

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: PC12, Ls174T or Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)
  • Flow Cytometry (Not Suitable)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p21WAF1 Ab-6
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Human, Rat
IgG1
Immunofluorescence, Western Blot
HJ21; same as WA-1
Unconjugated
Human recombinant p21 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.