missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p21/CIP1/CDKN1A Antibody (CIP1/2275R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-80488-100UG
This item is not returnable.
View return policy
Description
p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| p21/CIP1/CDKN1A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 - 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-Page | |
| CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CIP1/2275R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CDKN1A | |
| Recombinant full-length human CDKN1A protein | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific | |
| 1026 | |
| Store at 2 - 8°C/ 1 year from date of receipt | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction