missing translation for 'onlineSavingsMsg'
Learn More

p21/CIP1/CDKN1A Antibody (CIP1/2275R), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18322660 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322660 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322660 Supplier Novus Biologicals Supplier No. NBP280489AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21/CIP1/CDKN1A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CIP1/2275R
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Host Species Rabbit
Immunogen Recombinant full-length human p21/CIP1/CDKN1A protein (Uniprot: P38936)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1026
Test Specificity This monoclonal antibody recognizes a 21kDa protein, identified as the p21WAF1 tumor suppressor protein. This monoclonal antibody is highly specific to p21 and shows no cross-reaction with other closely related mitotic inhibitors. p21WAF1 is a specific inhibitor of cdks and a tumor suppressor involved in the pathogenesis of a variety of malignancies. The expression of this gene acts as an inhibitor of the cell cycle during G1 phase and is tightly controlled by the tumor suppressor protein p53. Its expression is induced by the wild type, but not mutant, p53 suppressor protein. Normal cells generally display a rather intense nuclear p21 expression. Loss of p21 expression has been reported in many carcinomas (gastric carcinoma, non-small cell lung carcinoma, thyroid carcinoma).
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.