missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p21 ARC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114880
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARPC3.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been conserved through evolution and is implicated in the control of actin polymerization in cells. Alternatively spliced transcript variants have been found for this gene.
Specifications
| p21 ARC | |
| Polyclonal | |
| Unconjugated | |
| ARPC3 | |
| 1110006A04Rik; 21kDa; AA408672; actin related protein 2/3 complex subunit 3; actin related protein 2/3 complex subunit 3, 21kDa; actin related protein 2/3 complex, subunit 3; actin related protein 2/3 complex, subunit 3, 21kDa; actin-related protein 2/3 complex subunit 3; AI788639; ARC21; Arp2/3 complex 21 kDa subunit; Arp2/3 complex subunit p21-Arc; ARP2/3 protein complex subunit p21; Arpc3; p21-Ar; p21Arc; p21-Arc | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10094, 288669, 56378 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O15145, Q9JM76 | |
| ARPC3 | |
| A synthesized peptide derived from human ARPC3(Accession O15145), corresponding to amino acid residues A64-N114. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction