missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p19 INK4d Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527704
This item is not returnable.
View return policy
Description
Recommended positive controls: Rat bone marrow. Predicted reactivity: Mouse (88%), Rat (88%), Rhesus Monkey (98%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
p19 is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well as spermatogenesis.
Specifications
| p19 INK4d | |
| Polyclonal | |
| Unconjugated | |
| CDKN2D | |
| CDK inhibitor p19INK4d; CDK4I; CDKN2D; cell cycle inhibitor, Nur77 associating protein; cyclin dependent kinase inhibitor 2D; cyclin-dependent kinase 4 inhibitor D; cyclin-dependent kinase 4 inhibitor D p19; cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4); inhibitor of cyclin-dependent kinase 4d; INK4d; MTS1; p16-INK4; p16-INK4a; p19; p19INK4d; p19-INK4D; similar to cyclin-dependent kinase inhibitor 2D | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1032, 494444 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| P55273 | |
| CDKN2D | |
| Recombinant protein encompassing a sequence within the center region of human p19 INK4d. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction