missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p19 INK4d Polyclonal specifically detects p19 INK4d in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | p19 INK4d |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant p19 INK4d (P55273; Met10-Leu166). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?