missing translation for 'onlineSavingsMsg'
Learn More

p18INK4c/CDKN2C Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30734070 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734070 Supplier Novus Biologicals Supplier No. AF7746MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

p18INK4c/CDKN2C Polyclonal antibody specifically detects p18INK4c/CDKN2C in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen p18INK4c/CDKN2C
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CDK6 inhibitor p18, CDKN6, cyclin-dependent inhibitor, cyclin-dependent kinase 4 inhibitor C, Cyclin-dependent kinase 6 inhibitor, cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4), INK4Ccyclin-dependent kinase 6 inhibitor p18, p18, p18-INK4C, p18-INK6
Host Species Goat
Immunogen E. coli-derived recombinant human p18INK4c/CDKN2C, Ala2-Gln168, Accession # P42773
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1031
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.