missing translation for 'onlineSavingsMsg'
Learn More

p16INK4a/CDKN2A Antibody, FITC, Novus Biologicals™

Product Code. 18325282 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325282 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325282 Supplier Novus Biologicals Supplier No. NBP298881F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

p16INK4a/CDKN2A Polyclonal specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p16INK4a/CDKN2A
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human p16INK4a/CDKN2A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.