Learn More
P16-INK4A Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10883-100UL
Description
The P16-INK4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human P16-INK4A. This antibody recognizes human antigen. The P16-INK4A antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
| P16-INK4A | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 1029 | |
| IgG |