missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P15RS Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23652-1-AP-150UL
This item is not returnable.
View return policy
Description
P15RS is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15 and may have a role in cell cycle regulation (Liu et al., 2002 ).Specifications
| P15RS | |
| Polyclonal | |
| Unconjugated | |
| RPRD1A | |
| HsT3101, p15INK4B related protein, P15RS, RPRD1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225283, 55197 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VDS4, Q96P16 | |
| RPRD1A | |
| P15RS Fusion Protein Ag20409 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction