missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P15RS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35610-20ul
This item is not returnable.
View return policy
Description
P15RS Polyclonal antibody specifically detects P15RS in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| P15RS | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Cyclin-dependent kinase inhibitor 2B-related protein, Cyclin-dependent kinase inhibitor 2B-related protein (p15INK4B-related protein), FLJ10656, HsT3101, MGC19513, p15INK4B-related protein, P15RScyclin-dependent kinase 2B-inhibitor-related protein, regulation of nuclear pre-mRNA domain containing 1A, regulation of nuclear pre-mRNA domain-containing protein 1A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 47-276 of human P15RS (NP_001290341.1).,, Sequence:, PVIVEAFKHVSSETDESCKKHLGRVLSIWEERSVYENDVLEQLKQALYGDKKPRKRTYEQIKVDENENCSSLGSPSEPPQTLDLVRALQDLENAASGDAAVHQRIASLPVEVQEVSLLDKITDKESGERLSKMVEDACMLLADYNGRLAAEIDDRKQLTRMLADFLRCQKEALAEKEHKLEEYKRKLARVSLVRKELRSRIQSLPDLSRLPNVTGSHMHLPFAGDIYSED | |
| 20 μL | |
| Cell Cycle and Replication | |
| 55197 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction