missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p15INK4b/CDKN2B Antibody (OTI3B6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73184MFV500
This item is not returnable.
View return policy
Description
p15INK4b/CDKN2B Monoclonal antibody specifically detects p15INK4b/CDKN2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| p15INK4b/CDKN2B | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI3B6 | |
| Flow Cytometry | |
| CDK4I, cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4), cyclin-dependent kinases 4 and 6 binding protein, INK4BCDK4B inhibitor, MTS-2, MTS2CDK inhibitory protein, Multiple tumor suppressor 2, p14_CDK inhibitor, p14_INK4B, p14-INK4b, p15 CDK inhibitor, p15_INK4B, P15cyclin-dependent kinase 4 inhibitor B, p15INK4b, p15-INK4b, TP15 | |
| Full length human recombinant protein of human CDKN2B(NP_004927) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1030 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction