missing translation for 'onlineSavingsMsg'
Learn More

p15INK4b/CDKN2B Antibody (OTI3B6), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18407312 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407312 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407312 Supplier Novus Biologicals Supplier No. NBP273184MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p15INK4b/CDKN2B Monoclonal antibody specifically detects p15INK4b/CDKN2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen p15INK4b/CDKN2B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B6
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CDK4I, cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4), cyclin-dependent kinases 4 and 6 binding protein, INK4BCDK4B inhibitor, MTS-2, MTS2CDK inhibitory protein, Multiple tumor suppressor 2, p14_CDK inhibitor, p14_INK4B, p14-INK4b, p15 CDK inhibitor, p15_INK4B, P15cyclin-dependent kinase 4 inhibitor B, p15INK4b, p15-INK4b, TP15
Host Species Mouse
Immunogen Full length human recombinant protein of human CDKN2B(NP_004927) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1030
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.