missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p14ARF/CDKN2A Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-111MFV450
This item is not returnable.
View return policy
Description
p14ARF/CDKN2A Polyclonal antibody specifically detects p14ARF/CDKN2A in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| p14ARF/CDKN2A | |
| Polyclonal | |
| Flow Cytometry | |
| ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
| A synthetic peptide made to a portion of human p14ARF (between residues 50-150). [Swiss-Prot# Q8N726] | |
| 0.1 mL | |
| ABC Transporters, Biologically Active Proteins, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
| 1029 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction