missing translation for 'onlineSavingsMsg'
Learn More

p130Cas/BCAR1, p Tyr410 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18430575 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430575 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430575 Supplier Novus Biologicals Supplier No. NBP254638G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

p130Cas/BCAR1 Polyclonal antibody specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen p130Cas/BCAR1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas
Host Species Rabbit
Immunogen p130 Cas (phospho Tyr410) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 410 (G-V-Y (p)-A-V) derived from Human p130 Cas .
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9564
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.