missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p130Cas/BCAR1 Antibody (JE36-56), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32071
This item is not returnable.
View return policy
Description
p130Cas/BCAR1 Monoclonal antibody specifically detects p130Cas/BCAR1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| p130Cas/BCAR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
| Recombinant protein within Human p130Cas/BCAR1 aa 1-100/ 870. (Uniprot: P56945) | |
| 100 μL | |
| Phospho Specific | |
| 9564 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE36-56 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction