missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p130 Recombinant Rabbit Monoclonal Antibody (7H2L13)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703847
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Monkey, Mouse, Rat, Bovine.
RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
Specifications
| p130 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q08999 | |
| RBL2 | |
| Peptides corresponding to human p130 [1) aa175-aa192 2) aa1122-aa1139]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 7H2L13 | |
| Unconjugated | |
| RBL2 | |
| 130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RB transcriptional corepressor like 2; Rb2; rb2 p130; Rbl2; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130); retinoblastoma-like protein 2; Retinoblastoma-related protein 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction