missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582690
This item is not returnable.
View return policy
Description
Immunogen sequence: IAGSPLTPRR VTEVRADTGG LGRSITSPTT LYDRYSSPPA STTRRRLFVE NDSPSDGGTP GRMPPQPLVN AVPVQNVSGE TVSVTPVPGQ TLVTMATATV TANNGQTVTI PVQGIANENG GITFFPVQV Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 91%.
RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
Specifications
| p130 | |
| Polyclonal | |
| Unconjugated | |
| RBL2 | |
| 130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RB transcriptional corepressor like 2; Rb2; rb2 p130; Rbl2; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130); retinoblastoma-like protein 2; Retinoblastoma-related protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q08999 | |
| RBL2 | |
| Recombinant protein corresponding to Human RBL2. Recombinant protein control fragment (Product #RP-91613). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction