missing translation for 'onlineSavingsMsg'
Learn More

p130 Antibody (2H7), Novus Biologicals™

Product Code. 18372859 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372859 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372859 Supplier Novus Biologicals Supplier No. H00005934M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p130 Monoclonal antibody specifically detects p130 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen p130
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2H7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005602
Gene Alias FLJ26459, P130, PRB2, RB2, RBR-2, retinoblastoma-like 2 (p130), retinoblastoma-like protein 2,130 kDa retinoblastoma-associated protein, Retinoblastoma-related protein 2
Host Species Mouse
Immunogen RBL2 (NP_005602, 416 a.a. ~ 515 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VTPVSTATHSLSRLHTMLTGLRNAPSEKLEQILRTCSRDPTQAIANRLKEMFEIYSQHFQPDEDFSNCAKEIASKHFRFAEMLYYKVLESVIEQEQKRLG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5934
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.