missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p120-catenin Rabbit anti-Human, Clone: 4383R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | p120-catenin |
|---|---|
| Clone | 4383R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
p120-catenin Monoclonal specifically detects p120-catenin in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| p120-catenin | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 1500 | |
| IgG | |
| Protein A or G purified |
| 4383R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
| Synthetic peptide corresponding to p120-catenin residues within aa900-1000 of p120-catenin was used as an immunogen. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title