missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p120-catenin Antibody (M354), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23202
This item is not returnable.
View return policy
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| p120-catenin | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| M354 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:50 | |
| Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
| Clone (M354) was generated from a peptide that includes amino acids from the central region of human p120-catenin. This peptide sequence is highly conserved in rat and mouse p120-catenin. [UniProt# O60716] | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 1500 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction