missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
Specifications
| Antigen | p120-catenin |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Clone | 25a |
| Conjugate | DyLight 755 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN |
| Host Species | Mouse |
| Immunogen | Mouse p120-catenin (pY96) (Uniprot: O60716) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?