missing translation for 'onlineSavingsMsg'
Learn More

p115, USO1 Mouse anti-Human, Mouse, Rat, Clone: 3B7D8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68100-1-IG-20UL

 View more versions of this product

Product Code. 17721149

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

p115, USO1 Monoclonal antibody specifically detects p115, USO1 in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISA
Specifications

Specifications

p115, USO1
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Mouse
20 μL
8615
Store at -20°C
Liquid
Immunohistochemistry, Western Blot, ELISA
3B7D8
Unconjugated
BC022847
Protein A purification
Primary
Human, Mouse, Rat
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.