missing translation for 'onlineSavingsMsg'
Learn More

p107 Antibody (1A5), Novus Biologicals™

Product Code. 18369819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369819 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369819 Supplier Novus Biologicals Supplier No. H00005933M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p107 Monoclonal antibody specifically detects p107 in Human samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen p107
Applications Western Blot, ELISA, Proximity Ligation Assay
Classification Monoclonal
Clone 1A5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Proximity Ligation Assay
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32247
Gene Alias cellular protein 107,107 kDa retinoblastoma-associated protein, cp107, p107MGC40006, PRB1, retinoblastoma-like 1 (p107), retinoblastoma-like protein 1
Host Species Mouse
Immunogen RBL1 (AAH32247, 905 a.a. ~ 1014 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IDCDLEDATKTPDCSSGPVKEERGDLIKFYNTIYVGRVKSFALKYDLANQDHMMDAPPLSPFPHIKQQPGSPRRISQQHSIYISPHKNGSGLTPRSALLYKFNGSPSKVR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5933
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.