Learn More
MEDICOM Kolmi™ Vitals Type II Earloop Mask
Product Code. 31781426 Shop All Medicom Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Pack of 960
Product Code. Size Unit Size
31781426 One Size Pack of 960
1 options
Product Code. 31781426 Supplier MEDICOM Supplier No. M1310140
  • ONLINE EXCLUSIVE PRICE
    £72.35
    £61.83 / Pack of 960
    Save £10.52
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Kolmi™ Vitals Type II mask: protects others from droplets; high filtration and comfortable fit thanks to Origami folding.
  • Protects the environment from droplets emitted by the wearer
  • Effective against bacterias, particles and viruses (BFE, PFE, and VFE tested)
  • Kolmi masks feature the exclusive Origami folding and Smile welding for greater comfort and an ideal fit on the wearer's face
  • Hypoallergenic
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Earloops
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated