Learn More
MEDICOM Kolmi™ Feeldry Type II Tie-On Mask
Product Code. 31781325 Shop All Medicom Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Pack of 360
Product Code. Size Unit Size
31781325 One Size Pack of 360
1 options
Product Code. 31781325 Supplier MEDICOM Supplier No. M2410140
  • ONLINE EXCLUSIVE PRICE
    £49.23
    £42.09 / Pack of 360
    Save £7.14
    14% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Kolmi™ Feeldry Type II mask: protects others from droplets with high filtration and durable comfort thanks to Feeldry technology.
  • Protects the environment from droplets emitted by the wearer
  • Effective against bacteria, particles and viruses (BFE, PFE, and VFE)
  • Kolmi masks feature the exclusive Origami folding for greater comfort and an ideal fit on the wearer's face
  • Featuring a wide and ultra-flexible nose strip, they ensure an excellent fit to the face and effectively limit leakage around the nose, thereby significantly reducing fogging
  • For maximum comfort, the inner layer is designed with a material that limits irritation to the face
  • With Feeldry technology the inner layer has been specially designed to absorb perspiration and avoid the feeling of humidity inside the mask
  • Hypoallergenic
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Ties
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated