Learn More
MEDICOM Kolmi™ Vitals Type IIR Tie-On Mask
Product Code. 31781301 Shop All Medicom Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Pack of 300
Product Code. Size Unit Size
31781301 One Size Pack of 300
1 options
Product Code. 31781301 Supplier MEDICOM Supplier No. M3310140
  • ONLINE EXCLUSIVE PRICE
    £34.60
    £29.57 / Pack of 300
    Save £5.03
    14% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Kolmi™ Vitals Type IIR mask: protects others and wearer from biological fluid splashes with a reliable fit.
  • Protects the environment from droplets emitted by the wearer while providing protection from splashes of biological fluids
  • Effective against bacterias, particles and viruses (BFE, PFE, and VFE tested)
  • Kolmi masks feature the exclusive Origami folding for greater comfort and an ideal fit on the wearer's face
  • Featuring a wide and ultra-flexible nose strip, they ensure an excellent fit to the face and effectively limit leakage around the nose, thereby significantly reducing fogging
  • Hypoallergenic
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Ties
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated