Learn More
Medicom™ WeePro Type PB 6 Overshoe with Anti-slip Sole
Product Code. 17988300 Shop All Medicom Products
Change view
Click to view available options
Size:
Large/X-Large
Unit Size:
Pack of 400
Product Code. Size Unit Size
17988300 Large/X-Large Pack of 400
1 options
Product Code. 17988300 Supplier Medicom™ Supplier No. WLCCE/ADLXL
  • ONLINE EXCLUSIVE PRICE
    £344.31
    £297.78 / Pack of 400
    Save £46.53
    13% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WeePro Type PB 6 Overshoe with Anti-slip Sole has strong anti-slip power and is made of a very low-lint material.
  • Lint-free material to avoid any contamination in a controlled critical environment
  • Thick PVC sole with a high anti-slip property over dry or wet floors
  • Elastic wrap at ankle and heel to keep the overshoes straight
  • Anti-static material to allow easy dissipation of electrostatic charges
  • Ideal protection against low concentration chemical splashes, powder, adhesives, fiberglass, solvents, paints
Specifications
Size Large/X-Large
Material Microporous Fabric
Latex Free No
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Resistance Light Chemical Splash
Product Type Controlled Environments Shoe Covers