missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ One Type IIR Medical Mask with Visor

Product Code. 17790199 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Honey
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17790199 Blue Pack of 25
17740209 Honey Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17790199 Supplier Kolmi Hopen™ Supplier No. M34101V30
  • ONLINE EXCLUSIVE PRICE
    £34.45
    £30.41 / Pack of 25
    Save £4.04
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The built-in visor of this medical mask helps protect the wearer from being splashed by biological fluids.

  • Protection against human liquids, bacteria, particles, and viruses
  • Anatomical folding reduces exposure to face
  • Inner layer designed to wick away moisture and sweat
  • Transparent polyester visor with anti-fog treatment on both sides

Specifications

Color Blue
Fastening Type Ties
Material Polypropylene
Style Pleated with Eyeshield
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485
Disposable Single-use
For Use With (Application) Medical
Latex Free Yes
Packaging Quantity 25 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.