missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Elasticated Helmet Hood with Integrated Mask

Product Code. 17770139 Shop All Medicom Products
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17770139 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17770139 Supplier Kolmi Hopen™ Supplier No. 70.702
  • ONLINE EXCLUSIVE PRICE
    £11.72
    £9.71 / Pack of 50
    Save £2.01
    17% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This elasticated helmet hood with face mask provides a high level of protection by leaving only the eyes exposed.

  • Ultra-effective protection for the wearer and the environment
  • Integrated, 2-ply pleated mask
  • Available for color code tracking
  • 25 g/m2 fabric weight

Specifications

Material Polypropylene
Color Blue
Disposable Single-use
Design Type Helmet
Quantity 50 Pk.
Includes Hoods and Polybags
For Use With (Application) Industrial, Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.