missing translation for 'onlineSavingsMsg'
Learn More

Kolmi™ Op Air Pro Oxygen FFP2 NR D Mass Balance Mask

Product Code. 17764789 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17764789 Medium Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17764789 Supplier Kolmi™ Supplier No. M52014WHMB
  • ONLINE EXCLUSIVE PRICE
    £19.87
    £15.25 / Pack of 50
    Save £4.62
    23% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Op Air Pro Oxygen FFP2 mask is easy to put on with high breathability due to high quality raw material. Made with the Mass Balance approach from recycled materials.

  • Filtering half mask, efficiency class FFP2 NR D developed to protect against solid and liquid aerosols and particles.
  • Type IIR certified, it protects the environment against droplets emitted by the wearer and it protects him from potential body fluids splashes.
  • Knitted headloops ultrasonically welded for an optimal comfort.
  • Thanks to its shape, and the use of soft materials, the OP-air Pro Oxygen fits perfectly to the face to limit inward leakage.
  • This mask is hypoallergenic and has been dermatologically tested.
  • OP-air Pro Oxygen is made in France.

Specifications

Certifications/Compliance EN 149: 2001 + A1: 2009, DM Class I, EN 14683: 2019 + AC: 2019
Color White
Size Medium
For Use With (Application) Industry
Style Pouch
Strap Attachment Type Headband
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.