missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ Type II Medical Masks with Earloops

Product Code. 17760179 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 2000
Pack of 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17760179 Blue Pack of 2000
17770179 Green Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17760179 Supplier Kolmi Hopen™ Supplier No. M14311302000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pleated medical mask with anti-fog nose clip can be used during transport of contagious patients.

  • High filtration media (polypropylene)
  • Two wide and comfortable elastic bands
  • Very soft polypropylene edges
  • Anatomical folding limits exposure to face
  • Integrated anti-fog nose bar
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)

Specifications

Color Blue
Fastening Type Earloops
Material Polypropylene
Style Pleated
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683, ISO 13485, EN ISO 14971
Disposable Single-use
For Use With (Application) Medical
Packaging Quantity 2000 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.