missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ Type II Surgical Masks with Earloops

Product Code. 17750179 Shop All Medicom Products
Change view
Click to view available options
Type II Op-Air Surgical Masks with Earloops; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Fastening Type: Earloops; Disposable: Yes; Style: Pleated; Color: White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17750179 Type II Op-Air Surgical Masks with Earloops; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Fastening Type: Earloops; Disposable: Yes; Style: Pleated; Color: White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17750179 Supplier Kolmi Hopen™ Supplier No. M1430130
  • ONLINE EXCLUSIVE PRICE
    £6.13
    £5.18 / Pack of 60
    Save £0.95
    15% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pleated medical mask with anti-fog nose clip for healthcare workers during surgical intervention.

  • High filtration media (polypropylene)
  • Two wide and comfortable elastic bands
  • Very soft polypropylene edges
  • Anatomical folding limits exposure to face
  • Integrated nose bar
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)

Specifications

Color White
Fastening Type Earloops
Material Polypropylene
Style Pleated
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683, ISO 13485, ISO 9001
Disposable Single-use
For Use With (Application) Medical
Packaging Quantity 60 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.