missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ INOX™ Premium Clip Cap Dispenser

Product Code. 17750099 Shop All Medicom Products
Change view
Click to view available options
Premium Clip Cap Dispenser; Color: White; Material: Stainless Steel; Depth (Metric): 200 mm; Height (Metric): 400 mm; Width (Metric): 200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17750099 Premium Clip Cap Dispenser; Color: White; Material: Stainless Steel; Depth (Metric): 200 mm; Height (Metric): 400 mm; Width (Metric): 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17750099 Supplier Kolmi Hopen™ Supplier No. 20.B1050
  • ONLINE EXCLUSIVE PRICE
    £233.24
    £201.02 / Pack of 1
    Save £32.22
    13% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premium dispenser for caps of all types of bulk styling, beard covers, overshoes, overboots, cuffs, and more.

  • Complies with the HACCP approach
  • Suitable for all brands
  • Easy cleaning
  • Wall mounted (screws and dowels supplied)

Specifications

Color Silver
Depth (Metric) 200 mm
Width (Metric) 200 mm
Height (Metric) 400 mm
Material Stainless Steel
For Use With (Application) Medical, Dental, Industrial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.