missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Polypropylene Bouffant Caps, 53 cm Diameter

Product Code. 17740129 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Green
White
Includes:
Caps and Polybags
Caps and Wall Dispenser
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Includes Unit Size
17740129 White Caps and Wall Dispenser Pack of 100
17750129 White Caps and Polybags Pack of 100
17760129 Blue Caps and Wall Dispenser Pack of 100
17770129 Green Caps and Wall Dispenser Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17740129 Supplier Kolmi Hopen™ Supplier No. 70.011
  • ONLINE EXCLUSIVE PRICE
    £8.00
    £4.62 / Pack of 100
    Save £3.38
    42% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

These comfortable bouffant caps provide additional hygiene control for various industrial environments and come in polybags or wall dispensers for convenient use.

  • Lightweight for improved comfort
  • Ability to use a color code for tracking
  • Adapts to different head sizes
  • Single elastic band
  • 12 g/m2 material weight

Specifications

Material Polypropylene
Color White
Disposable Single-use
Design Type Bouffant
Sterility Non-sterile
Certifications/Compliance EN ISO 9001
Quantity 100 Pk.
Diameter (Metric) 53 cm
Includes Caps and Wall Dispenser
For Use With (Application) Industrial, Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.