missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Elasticated SMS Gown with Collar and Zipper

Product Code. 17740119 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17740119 Large Pack of 50
17750119 X-Large Pack of 50
17760119 2X-Large Pack of 50
17770119 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17740119 Supplier Kolmi Hopen™ Supplier No. 40.507/03
  • ONLINE EXCLUSIVE PRICE
    £135.96
    £128.15 / Pack of 50
    Save £7.81
    5% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

The polyethylene film of these gowns makes it possible to reduce the particulate release of fibers in the industrial workplace.

  • Single use, anti-static hygiene gown provides the user with full body protection
  • Protects against activity related soiling and contamination
  • Microporous, polyethylene lining is comfortable, and waterproof

Specifications

Product Type Hygiene Gown
Size Large
Gender Unisex
Material Polypropylene, SMS, Microporous Polyethylene Laminate
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Certifications/Compliance (Europe) EN 1149-5
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Weight (Metric) 65 g/m2
No. per Pack 50 Pk.
Construction Made in China
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.