missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Peaked Bouffant Cap for Cropped Hair with Dispenser Box

Product Code. 17730139 Shop All Medicom Products
Change view
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17730139 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17730139 Supplier Kolmi Hopen™ Supplier No. 70.301
  • ONLINE EXCLUSIVE PRICE
    £17.04
    £14.16 / Pack of 100
    Save £2.88
    16% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This comfortable bouffant cap is ideal for keeping short hair concealed for working in industrial environments.

  • Lightweight and comfortable
  • Not suitable for workers with long hair
  • 37 g/m2 fabric weight

Specifications

Material Polypropylene
Color White
Disposable Single-use
Design Type Peaked Cap
Sterility Non-sterile
Quantity 100 Pk.
Length (Metric) 28 cm (Cap), 7 cm (Visor)
Includes Caps and Dispenser Box
For Use With (Application) Industrial, Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.