missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ Type IIR Splash-resistant Medical Mask with Head Straps

Product Code. 17710219 Shop All Medicom Products
Change view
Click to view available options
Op-Air Type IIR Splash-resistant Medical Mask with Head Straps; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683:2019+AC:2019; Color: White; Disposable: Yes; Fastening Type: Elastic Straps; Material: Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17710219 Op-Air Type IIR Splash-resistant Medical Mask with Head Straps; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683:2019+AC:2019; Color: White; Disposable: Yes; Fastening Type: Elastic Straps; Material: Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17710219 Supplier Kolmi Hopen™ Supplier No. M3900130
  • ONLINE EXCLUSIVE PRICE
    £46.86
    £20.47 / Pack of 50
    Save £26.39
    56% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Elastic, over-the-head straps make this medical mask extra secure while protecting the wearer from splashes.

  • Resistant to liquid projections with pressure levels up to 16 kPa
  • Soft head loops for maximum comfort and security
  • Anatomical folding reduces exposure to face
  • Integrated nose piece
  • Fiberglass-free filter
  • Production in ISO 8 clean room (particle and microbiological control)
  • Individually packed to ensure better cleanliness of the mask

Specifications

Color White
Fastening Type Elastic Straps
Material Polypropylene
Dimensions (L x W) 230 x 100 mm
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683:2019+AC:2019
Disposable Single-use
For Use With (Application) Medical
Latex Free Yes
Packaging Quantity 50 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.