missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ SafeFeet Polypropylene Overshoes with SkidGuard
GREENER_CHOICE

Product Code. 17700159 Shop All Medicom Products
Change view
Click to view available options
Color:
White
White, Green
White, Pink
Size:
X-Large
2X-Large
Unit Size:
Pack of 300
Pack of 400
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size unitSize
17700159 White 2X-Large Pack of 300
17780149 White, Pink X-Large Pack of 400
17790149 White, Green X-Large Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17700159 Supplier Kolmi Hopen™ Supplier No. H07101A
  • ONLINE EXCLUSIVE PRICE
    £81.11
    £78.84 / Pack of 300
    Save £2.27
    2% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Help prevent falls in the workplace with these skid-resistant, polypropylene overshoes.

  • Waterproof and non-slip soles
  • Breathable and lightweight
  • Elastic ankle support
  • Non-woven polypropylene with polyethylene soles
  • Made in France

Specifications

Size 2X-Large
Material Polypropylene
Color White
Disposable Single-use
Thickness 60 μm
For Use With (Application) Medical, Industrial
Certifications/Compliance ISO 13485, (EU) 2017/745
Packaging Quantity 300 Pk.
Weight (Metric) 34 g/m2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.