missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ SMS Isolation Gown with Thumb Holes
GREENER_CHOICE

Product Code. 17700149 Shop All Medicom Products
Change view
Click to view available options
SMS Isolation Gown with Thumb Holes; Color: Yellow; Disposable: Yes; Material: SMS; For Use With (Application): Medical; Size: X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17700149 SMS Isolation Gown with Thumb Holes; Color: Yellow; Disposable: Yes; Material: SMS; For Use With (Application): Medical; Size: X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17700149 Supplier Kolmi Hopen™ Supplier No. A69962
Add to basket
Add to basket

The three-ply protective fabric of this gown combines strength and resistance to fluids with softness and comfort, thanks to its breathability.

  • High quality infection and fluid protection
  • Consists of a layer of polypropylene obtained by extrusion-blow molding inserted between two layers of polypropylene obtained by spinning-topping
  • Large opening for passage of the head
  • Patented 'Flexneck' technology consisting of double elastic
  • Long sleeves with thumb holes that completely cover wrists
  • Designed to protect healthcare professionals and patients from cross infection
  • To be worn over traditional clothing

Specifications

Size X-Large
For Use With (Application) Medical
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material SMS
Color Yellow
Disposable Single-use
Sterility Non-sterile
Weight (Metric) >23.5 g/m2
Autoclavable Not Autoclavable
No. per Pack 10 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.