missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCP-3 ProQuantum Immunoassay Kit
GREENER_CHOICE

Product Code. 17113119
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
96 reactions
Each
This item is not returnable. View return policy

Product Code. 17113119

Brand: Invitrogen™ A356005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

This is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80098
Assay Range 0.064 to 5000 pg/mL
Assay Sensitivity <0.05 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Protein Quantitation
For Use With (Equipment) qPCR Instruments
Gene ID (Entrez) 6354
Gene Symbol FIC; MARC; MCP-3; MCP3; NC28; SCYA6; SCYA7
Incubation Time 1 hr.
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Storage Requirements Frozen at -20°C
Target CCL7 (MCP3)
Test Time 30 mins
Total Assay Time 2 hrs
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.