missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Entris™ II Advanced Analytical Balances

Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs

Brand:  Sartorius BCA324I-1S


3553.00 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 4.60000kg

 View more versions of this product

Product Code. 16646253

  • £3740.00 / Each
Estimated Shipment: 19-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • Accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications

Specifications

CE Certificate, CSA for Canada
With Internal Calibration
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤1.5 sec.
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
320 g
0.1 mg
Electronic Analytical Balance
100/240 V (±10%)
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers